Triethylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:121-44-8
:
Formula:(C₂H₅)₃N
MDL Number:MFCD00009051
:101.19 g/mol
Merck Index:13,09740
Boiling Pt:90 °C (1013 hPa)
Melting Pt:–115 °C
Density:0.729 g/cm³ (20 °C)
EINECS:204-469-4
UN:1296
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:–11 °C
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient