Order Entry
Germany
ContactUsLinkComponent
 
N,N-Dimethylformamide di-tert-butyl acetal, Supelco®
  395005-25ML
 :  Merck
 :  
undefined
N,N-Dimethylformamide di-tert-butyl acetal, Supelco®
  395005-25ML
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • APPEARANCE (COLOR):
    Colorless to Yellow
  • APPEARANCE (FORM):
    Liquid
  • PURITY (GC AREA %):
    ≥ 90.0 %
  • PROTON NMR SPECTRUM:
    CONFORMS TO STRUCTURE

 

 

 :
 :203.32 g/mol
 :56…57 °C (8 mmHg) (lit.)
 :0.848 g/mL (25 °C) (lit.)
 :M-Clarity™ quality level = MQ100
 :253-222-7
 :GHS02
 :GHS07
 :36805-97-7
 :C₁₁H₂₅NO₂
 :MFCD00015002