Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Iodoacetamide
Iodoacetamide
Iodoacetamide
:
Serva
Iodoacetamide
:
Serva
:
144-48-9
:
:
ICH₂CONH₂
:
MFCD00008028
:
184.96 g/mol
:
266 °C (1013 hPa)
:
91…94 °C
:
Refrigerator
:
205-630-1
:
2811
:
6.1,III
:
GHS07