Iodoacetamide for synthesis, Sigma-Aldrich®
: Merck
:144-48-9
:
:ICH₂CONH₂
:MFCD00008028
:184.96 g/mol
:266 °C (1013 hPa)
:91…94 °C
:Refrigerator
:205-630-1
:2811
:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
- Solubility:(20 °C) soluble
- Melting Point:91 - 94 °C
- Molar Mass:184.96 g/mol
- Hill Formula:C2H4INO
- Chemical Formula:ICH2CONH2