ContactUsLinkComponent
Iodoacetamide
  786-228
 :  
Iodoacetamide
  786-228
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173
 :
 :184.96 g/mol
 :266 °C (1013 hPa)
 :91…94 °C
 :Refrigerator
 :205-630-1
 :2811
 :6.1,III
 :GHS07
 :144-48-9
 :ICH₂CONH₂
 :MFCD00008028