Iodoacetamide
Catalog # FLUO001085-25G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:184.96 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:91…94 °C
Storage Temperature:Refrigerator
EINECS:205-630-1
UN:2811
ADR:6.1,III
:GHS07
CAS Number:144-48-9
Formula:ICH₂CONH₂
MDL Number:MFCD00008028