Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Difluoroethanol
2,2-Difluoroethanol
2,2-Difluoroethanol
2,2-Difluoroethanol
Supplier:
FLUOROCHEM
Certificates
Expand image
2,2-Difluoroethanol
Supplier:
FLUOROCHEM
About this item
CAS Number:
359-13-7
:
Formula:
C₂H₄F₂O
MDL Number:
MFCD00042248
:
82.05 g/mol
Boiling Pt:
95 °C (1013 hPa)
Melting Pt:
–28,2 °C
Density:
1,296 g/cm³ (20 °C)
UN:
1987
ADR:
3,II