
2,2,3,3,3-Pentafluoro-1-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:422-05-9
:
Formula:C₃H₃F₅O
MDL Number:MFCD00004673
:150.05 g/mol
Boiling Pt:78…80 °C (1013 hPa)
Density:1.5 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:207-012-7
:GHS07
Frequently Bought Together