

1,1,1,3,3,3-Hexafluoro-2-propanol
Supplier: FLUOROCHEM
Certificates
About this item
CAS Number:920-66-1
:
Formula:C₃H₂F₆O
MDL Number:MFCD00011651
Merck Index:13,09598
Boiling Pt:59.1 °C (1013 hPa)
Melting Pt:–96 °C
Density:1.492 g/cm³ (20 °C)
Flash Pt:>100 °C
UN:1760
ADR:8,II
:168.04 g/mol
EINECS:213-059-4
:GHS05
:GHS07
Storage Temperature:Ambient