Trifluoroacetic anhydride, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:407-25-0
:
Formula:C₄F₆O₃
MDL Number:MFCD00000416
:210.03 g/mol
Boiling Pt:39.4 °C (1013 hPa)
Melting Pt:–65 °C
Density:1.487 g/cm³ (20 °C)
EINECS:206-982-9
ADR:8,I
:M-Clarity™ quality level = MQ100
UN:3265
:GHS05
:GHS07