Trifluoroacetic anhydride
Catalog # MOLEM27660426
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:210.03 g/mol
Boiling Pt:39.4 °C (1013 hPa)
Boiling Pt:39,4 °C (1013 hPa)
Melting Pt:–65 °C
Density:1,487 g/cm³ (20 °C)
EINECS:206-982-9
UN:3265
ADR:8,I
:GHS05
:GHS07
CAS Number:407-25-0
Formula:C₄F₆O₃
MDL Number:MFCD00000416