Trifluoroacetamide 97%
:354-38-1
:
:C₂H₂F₃NO
:MFCD00008008
:113.04 g/mol
:162…164 °C (760 mmHg)
:59…70 °C
:1 g/cm³ (20 °C)
:Ambient
:206-559-9
:GHS07
- Appearance:White to light yellow to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:68 to 75 °C
- GC:≥96.0 %