Order Entry
Germany
Orders LinkContactUsLinkComponent
 

EDTA tetrasodium salt ≥83%, TECHNICAL

Plastic bottle for solids, 1 kg
Catalog # 20299.291
Irritant
Corrosive
Supplier:   VWR Chemicals
EDTA tetrasodium salt ≥83%, TECHNICAL
Catalog # 20299.291
Supplier:  VWR Chemicals
CAS Number:  64-02-8
EDTA tetrasodium salt ≥83%, TECHNICAL
EDTA tetrasodium salt ≥83%, TECHNICAL

Plastic bottle for solids, 1 kg

CAS Number:  64-02-8
Price
156,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ethylenediaminetetraacetic acid tetrasodium salt, Ethylenedinitrilotetra-acetic acid tetrasodium salt, Tetrasodium ethylenediaminetetraacetate, EDTA tetrasodium, Edetic acid tetrasodium salt, Kestranal® 4S
  • CAS Number:64-02-8
  • Formula:C₁₀H₁₂N₂Na₄O₈
  • MDL Number:MFCD00150027
  • Molecular Weight:380.17 g/mol
  • Merck Index:12,03557
  • Melting Pt:> 300 °C
  • Density:6.9 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-573-9
Specifications

Specification Test Results

  • Assay:
    Min. 83 %

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg

Specifications