Order Entry
Germany
ContactUsLinkComponent
EDTA (ethylenediamine tetraacetic acid) ≥98%, TECHNICAL
Irritant
EDTA (ethylenediamine tetraacetic acid) ≥98%, TECHNICAL
Catalog # 20294.294
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
EDTA (ethylenediamine tetraacetic acid) ≥98%, TECHNICAL
Catalog # 20294.294
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

About this item

 :
 :292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
 :GHS07
CAS Number:60-00-4
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541