
Titriplex® II (EDTA (Ethylenediamine tetraacetic acid)) 99.4-100.6% (by complexometric analysis) ACS, Reag. Ph. Eur., Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
Specification Test Results
- Assay (complexometric):99.4 - 100.6 %
- Identity (IR-spectrum):conforms
- Appearance:White to almost white, fine crystalline to crystalline powder.
- in ammonia insoluble components:≤ 0.005 %
- Heavy metals (as Pb):≤ 10 ppm
- Ca (Calcium):≤ 10 ppm
- Fe (Iron):≤ 10 ppm
- Mg (Magnesium):≤ 5 ppm
- Nitrilotriacetic acid:≤ 0.1 %
- Sulfated ash:≤ 0.1 %
- Water (according to Karl Fischer):≤ 0.2 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.08417.0100_TDS_EN ProductPage_108417
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_108417