EDTA (ethylenediamine tetraacetic acid) free acid ≥99.5%, Ultrapure
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:60-00-4
:
Formula:C₁₀H₁₆N₂O₈
MDL Number:MFCD00003541
:292.25 g/mol
Merck Index:13,03546
Boiling Pt:614 °C (1013 hPa)
Melting Pt:220 °C
Density:0.86 g/cm³ (20 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:200-449-4
REACH:01-2119486399-18
:GHS07
Specification Test Results
- DNase:none detected
- Heavy Metals (as Pb):≤0.001%
- Insolubles (5%, 1N NaOH):Pass
- Iron:≤0.005%
- Magnesium:≤0.0005%
- Nitrilotriacetic Acid:≤0.1%
- pH (5%, Water) @25 °C:2.5 – 3.5
- Protease:none detected
- Purity:≥99.5%
- Residue after Ignition:≤0.2%
- RNase:none detected