Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
EDTA (Ethylenediamine tetraacetic acid)
EDTA (ethylenediamine tetraacetic acid) 99%
EDTA (ethylenediamine tetraacetic acid) 99%
:
Thermo Fisher Scientific
EDTA (ethylenediamine tetraacetic acid) 99%
:
Thermo Fisher Scientific
:
60-00-4
:
:
C₁₀H₁₆N₂O₈
:
MFCD00003541
:
292.25 g/mol
:
14,3517
:
614 °C (1013 hPa)
:
∼ 245 °C (dec.)
:
0.86 g/cm³ (20 °C)
:
>100 °C
:
Ambient
:
200-449-4
:
01-2119486399-18
:
GHS07
msds_ALFAA10713
Thermo Scientific Acroseal 25 Years