Azelaic acid
PROOMM0687.00
:
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
:
:188.22 g/mol
:13,00908
:371 °C (1013 hPa)
:106 °C
:1.0287 g/cm³ (20 °C)
:215 °C
:Ambient
:204-669-1
:GHS07
:123-99-9
:C₉H₁₆O₄
:MFCD00004432