Order Entry
Germany
ContactUsLinkComponent
Irritant
Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
Azelaic acid ≥90% (by GC), Technical Grade for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    2.4 g/l (20 °C)
  • Melting Point:
    107 °C
  • Molar Mass:
    188.22 g/mol
  • Boiling Point:
    500 °C (1013 hPa)
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    215 °C
  • Density:
    1.029 g/cm3 (20 °C)
  • pH:
    3.5 (1 g/l, H2O)
  • Synonyms:
    Nonanedioic acid
  • Hill Formula:
    C9H16O4
  • Chemical Formula:
    HOOC(CH2)7COOH

Specification Test Results

Related Information