Azelaic acid
Catalog # EHERC14126200
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:188.22 g/mol
Merck Index:13,00908
Boiling Pt:371 °C (1013 hPa)
Melting Pt:106 °C
Density:1.0287 g/cm³ (20 °C)
Flash Pt:215 °C
Storage Temperature:Ambient
EINECS:204-669-1
:GHS07
CAS Number:123-99-9
Formula:C₉H₁₆O₄
MDL Number:MFCD00004432