Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Azelaic acid
Azelaic acid tech. 80%, Technical Grade
Azelaic acid tech. 80%, Technical Grade
:
Thermo Fisher Scientific
Azelaic acid tech. 80%, Technical Grade
:
Thermo Fisher Scientific
:
123-99-9
:
:
C₉H₁₆O₄
:
MFCD00004432
:
188.22 g/mol
:
14,905
:
286 °C (100 mmHg)
:
∼ 102 °C
:
1.225
:
215 °C (419 °F)
:
204-669-1
:
GHS07
:
Ambient
msds_ALFAA17271
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years