Azelaic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:123-99-9
:
Formula:C₉H₁₆O₄
MDL Number:MFCD00004432
:188.22 g/mol
Merck Index:14,905
Boiling Pt:286 °C (100 mmHg)
Melting Pt:∼ 102 °C
Density:1.225
Flash Pt:215 °C (419 °F)
EINECS:204-669-1
:GHS07
Storage Temperature:Ambient