
Adipic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:124-04-9
:
Formula:HOOC(CH₂)₄COOH
MDL Number:MFCD00004420
Merck Index:13,00163
Boiling Pt:337 °C (1013 hPa)
Melting Pt:151 °C
Density:1.36 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:196 °C (closed cup)
:146.14 g/mol
EINECS:204-673-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:15 g/l (20 °C)
- Melting Point:151 - 154 °C
- Molar Mass:146.14 g/mol
- Bulk Density:700 kg/m3
- Boiling Point:330 °C (1013 hPa)
- Vapor Pressure:0.097 hPa (18.5 °C)
- Flash Point:196 °C
- Density:1.360 g/cm3 (25 °C)
- pH:2.7 (24.9 g/l, H2O, 25 °C)
- Ignition Point:405 °C
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_818650
SDS
ProductPage_818650 ProductPage_818650
Product specification
Frequently Bought Together