Order Entry
Germany
Orders LinkContactUsLinkComponent
Glutaric acid

Glutaric acid

Glass bottle for solids, 500 g
Catalog # MOLE30858763-500G
Irritant
Marketsource
Supplier:   Molekula
Glutaric acid
Catalog # MOLE30858763-500G
Supplier:  Molekula
Marketsource
CAS Number:  110-94-1
Glutaric acid

Glass bottle for solids, 500 g

CAS Number:  110-94-1
Price
Inquire For Price
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1,5-Pentanedioic acid
  • CAS Number:110-94-1
  • Formula:C₅H₈O₄
  • MDL Number:MFCD00004410
  • Molecular Weight:132.12 g/mol
  • Merck Index:13,04486
  • Boiling Pt:302 °C (1013 hPa)
  • Melting Pt:98 °C
  • Density:1.43 g/cm³ (15 °C)
  • Storage Temperature:Ambient
  • EINECS:203-817-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications