Order Entry
Germany
ContactUsLinkComponent
Sebacic acid 98%
Sebacic acid 98%
Certificates
undefined
Sebacic acid 98%

Specification Test Results

  • Appearance:
    White to off-white Powder or granules
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.3 %
  • GC:
    ≥97.5 %
  • Ash:
    ≤0.06 %

Specification Test Results

Related Information