Sarcosine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-97-1
:
Formula:C₃H₇NO₂
MDL Number:MFCD00004279
:89.09 g/mol
Merck Index:12,08519
Boiling Pt:185 °C (1013 hPa)
Melting Pt:63 °C
Density:1.301 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-538-6