Order Entry
Germany
ContactUsLinkComponent
 
Sarcosine for synthesis, Sigma-Aldrich®
 :  Merck
 
Sarcosine for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    1481 g/l (20°C)
  • Melting Point:
    208 °C
  • Molar Mass:
    600 kg/m3
  • Bulk Density:
    600 kg/m3
  • pH:
    6.1 (100 g/l, H2O, 20 °C)
  • Synonyms:
    Methylaminoacetic acid, N-Methylglycine
  • Hill Formula:
    C3H7NO2
  • Chemical Formula:
    CH3NHCH2COOH