Sarcosine 98%
:107-97-1
:
:C₃H₇NO₂
:MFCD00004279
:89.09 g/mol
:12,08519
:185 °C (1013 hPa)
:208 °C (dec.)
:1,301 g/cm³ (20 °C)
:>100 °C
:Ambient
:203-538-6
- Appearance:Colorless to white to yellow Powder or crystals
- Infrared spectrum:Conforms
- Water:≤2 %
- Solubility:(25 % in water) Clear colorless to light yellow
- Titration with HClO4:≥97.5 %