Order Entry
Germany
Orders LinkContactUsLinkComponent
L-Methionine

L-Methionine

Supplier:   MP Biomedicals
L-Methionine
Supplier:  MP Biomedicals
Certificates

4 Options Available Below

Product Details & Documents
Synonyms:  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  • CAS Number:63-68-3
  • Formula:C₅H₁₁NO₂S
  • MDL Number:MFCD00063097
  • Molecular Weight:149.21 g/mol
  • Merck Index:13,06004
  • Boiling Pt:278 °C (1013 hPa)
  • Melting Pt:136 °C
  • Density:1.373 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-562-9
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 27,00 €
    27,00 €
    1 * 25 g
       27,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 74,00 €
    74,00 €
    1 * 100 g
       74,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 308,00 €
    308,00 €
    1 * 500 g
       308,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 468,00 €
    468,00 €
    1 * 1 kg
       468,00 €