L-Methionine
Glass bottle for solids, 250 g
BACE4030850.0250
Marketsource
: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- :63-68-3
- :C₅H₁₁NO₂S
- :MFCD00063097
- :149.21 g/mol
- :13,06004
- :278 °C (1013 hPa)
- :136 °C
- :1.373 g/cm³ (20 °C)
- :Ambient
- :200-562-9
- Pack type:Glass bottle for solids
- Pk:250 g
- Cat. No.:BACE4030850.0250
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



