Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
D-Methionine
D-Methionine
D-Methionine
D-Methionine
Supplier:
BACHEM BIOCHEMICA
Certificates
Expand image
D-Methionine
Supplier:
BACHEM BIOCHEMICA
About this item
CAS Number:
348-67-4
:
Formula:
C₅H₁₁NO₂S
MDL Number:
MFCD00002622
:
149.21 g/mol
Merck Index:
13,06004
Boiling Pt:
278 °C (1013 hPa)
Melting Pt:
136 °C
EINECS:
206-483-6