Glycine
Catalog # UPTIUP018225
Supplier: Uptima
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1.64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-272-2
CAS Number:56-40-6
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131