Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycine
Glycine analytical reagent, Millipore®
Glycine analytical reagent, Millipore®
:
Merck
Glycine analytical reagent, Millipore®
:
Merck
:
56-40-6
:
:
NH₂CH₂COOH
:
MFCD00008131
:
75.07 g/mol
:
13,04504
:
181 °C (1013 hPa)
:
83 °C
:
1.64 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
Ambient
:
200-272-2
MERC1.04201.1000_TDS_eN
ProductPage_104201
msds_104201