Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Glycine
Glycine, Sigma-Aldrich®
Glycine, Sigma-Aldrich®
1.04169.1000
:
Merck
:
Glycine, Sigma-Aldrich®
1.04169.1000
:
Merck
:
Pack type:
Plastic bottle
Pk:
1 kg
Seq:
4024
:
:
75.07 g/mol
:
13,04504
:
181 °C (1013 hPa)
:
83 °C
:
1.64 g/cm³ (20 °C)
:
Ambient
:
200-272-2
:
56-40-6
:
NH₂CH₂COOH
:
MFCD00008131