Glycine ≥98%, powder
:56-40-6
:
:NH₂CH₂COOH
:MFCD00008131
:75.07 g/mol
:13,04504
:181 °C (1013 hPa)
:83 °C
:1.64 g/cm³ (20 °C)
:Ambient
:200-272-2
- Synonyms:Amino acetic acid; Gly; Amino ethanoic acid; Glycocoll
- Form:Powder
- Storage:Store at room temperature (15 to 30 °C)
- Isoelectric Point:6,06
- Loss on Drying:≤0,5%
- pka:for the α carboxyl group: 2,35; and for the α amino group: 9,78 at 25 °C