Order Entry
Germany
ContactUsLinkComponent
Glycine ≥99.5%, AnalaR NORMAPUR® analytical reagent
Glycine ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Glycine ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99,5 %
  • IR Spectrum:
    Passes test
  • pH (20°C; 5 %):
    5.9 - 6.3
  • Ignition residue (SO₄):
    Max. 0.05 %
  • Insolubility in water:
    Max. 30 ppm
  • Ninhydrin-positive substances:
    Max. 0.1 %
  • Cl (Chloride):
    Max. 70 ppm
  • NH₄ (Ammonium):
    Max. 0.02 %
  • SO₄ (Sulphate):
    Max. 25 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 5 ppm
  • Pb (Lead):
    Max. 1 ppm

Specification Test Results