Glycine ≥99.0%, Proteomics Grade
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1.64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Abs.@280 nm (1 M, Water):0.10
- Chloride:0.01%
- DNase:NONE
- Heavy Metals (as Pb):<0.002%
- Loss on Drying:0.2%
- pH (5 %, Water) @25 °C:5.9 – 6.4
- Protease:NONE
- Purity:99.0%
- RNase:NONE
- Solubility (10%, water):PASS
- Sulphate:0.05%
- Thin Layer Chromatography (TLC):ONE SPOT