Ammonium acetate
Catalog # SERA39750.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:77.08 g/mol
Merck Index:13,00495
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
Flash Pt:136 °C
EINECS:211-162-9
:GHS07
Storage Temperature:Ambient
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066