Order Entry
Germany
ContactUsLinkComponent
 
Potassium acetate ≥99.0% ACS, Fluka™
 
undefined
Potassium acetate ≥99.0% ACS, Fluka™

 

  • Assay (titration with Perchloric acid):
    Min. 99.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • pH (5 %, 25 °C):
    6.5 - 9.0
  • Calcium (Ca):
    Max. 0.005 %
  • Iron (Fe):
    Max. 5 ppm
  • Magnesium (Mg):
    Max. 0.002 %
  • Sodium (Na):
    Max. 0.03 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Chloride (Cl):
    Max. 0.003 %
  • Phosphate (PO₄):
    Max. 0.001 %
  • Sulfate (SO₄):
    Max. 0.002 %
  • IR spectrum:
    complying