
Diethyl malonate ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-53-3
:
Formula:C₇H₁₂O₄
MDL Number:MFCD00009195
:160.17 g/mol
Merck Index:13,03856
Boiling Pt:199 °C (1013 hPa)
Melting Pt:–50 °C
Density:1.055 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:93 °C
EINECS:203-305-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay (GC, area%):≥ 98.0 % (a/a)
- Density (d 20 °C/ 4 °C):1.053 - 1.057
- Identity (IR):passes test
- Boiling Point:199 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (40 °C)
- Flash Point:90 °C DIN 51758
- Refractive Index:1.4135 (20 °C, 589 nm)
- Explosion Limit:0.8 - 12.8 %(V)
- Density:1.055 g/cm3 (20 °C)
- Ignition Point:435 °C DIN 51794