Butyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:123-86-4
:
Formula:CH₃COO(CH₂)₃CH₃
MDL Number:MFCD00009445
:116.16 g/mol
Merck Index:13,01534
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–76 °C
Density:0.875 g/cm³ (20 °C)
EINECS:204-658-1
UN:1123
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:25 °C
:GHS02
:GHS07
Storage Temperature:Ambient