sec-Butyl acetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-46-4
:
Formula:C₆H₁₂O₂
MDL Number:MFCD00009328
:116.16 g/mol
Merck Index:13,01535
Boiling Pt:112 °C (1013 hPa)
Melting Pt:–98.9 °C
Density:0.87 g/cm³ (20 °C)
EINECS:203-300-1
UN:1123
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:19 °C
:GHS02