Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Paracetamol

Glass bottle for solids, 50 mg
Artikelnummer EDQMP0300000
Hersteller:   EDQM
Paracetamol
Artikelnummer EDQMP0300000
Hersteller:  EDQM
CAS-Nummer:  103-90-2
Paracetamol

Glass bottle for solids, 50 mg

CAS-Nummer:  103-90-2
Preis
270,00 €
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  Acetaminophen, N-Acetyl-4-aminophenol, 4′-Hydroxyacetanilide, 4'-Hydroxyacetanilide, Paracetamolum, 4-Acetamidophenol, N-(4-Hydroxyphenyl)acetamide, APAP
  • CAS Number:103-90-2
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00002328
  • Molecular Weight:151.16 g/mol
  • Merck Index:13,00048
  • Boiling Pt:388 °C (1013 hPa)
  • Melting Pt:168 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-157-5

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg

Specifications