Acetanilide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:103-84-4
:
Formula:C₈H₉NO
MDL Number:MFCD00008674
Merck Index:13,00051
Boiling Pt:304 °C (1013 hPa)
Melting Pt:115 °C
Density:1.22 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:174 °C
:135.17 g/mol
EINECS:203-150-7
:GHS07
Specification Test Results
- Solubility:5 g/l (20 °C)
- Melting Point:115 °C
- Molar Mass:135.16 g/mol
- Bulk Density:370 kg/m3
- Boiling Point:304 °C (1013 hPa)
- Vapor Pressure:0.002 hPa (20 °C)
- Flash Point:173 °C DIN 51758
- Density:1.22 g/cm3 (20 °C)
- pH:5 - 7 (10 g/l, H2O, 25 °C)
- Ignition Point:540 °C
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC8.22344.0500_TDS_EN ProductPage_822344 ProductPage_822344
Product specification
msds_822344