Order Entry
Germany
ContactUsLinkComponent
 
Urea 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 
undefined
Urea 99.5-100.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
 

 

  • Assay:
    99.5 - 100.5 %
  • Appearance of solution (10 %; water):
    Clear and colourless
  • IR Spectrum:
    Passes test
  • Acidity:
    Max. 0.0005 meq/g
  • Alkalinity:
    Max. 0.0005 meq/g
  • Melting point:
    132 - 135 °C
  • Biuret:
    Max. 0,1%
  • Heavy metals (as Pb):
    Max. 4 ppm
  • Ignition residue (SO4):
    Max. 50 ppm
  • Insolubility in water:
    Max. 30 ppm
  • Loss on drying (100-105°C):
    Max. 1.0 %
  • Cl (Chloride):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 10 ppm
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 2 ppm
  • Conforms to ACS:
    Passes test
  • Conforms to Reag. Ph.Eur.:
    Passes test