Order Entry
Germany
ContactUsLinkComponent
Urea ≥95%, GPR RECTAPUR®
Urea ≥95%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Urea ≥95%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 95 %
  • Melting point:
    132 - 135 °C
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Ignition residue (SO4):
    Max. 0.2 %
  • Cl (Chloride):
    Max. 5 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results