Order Entry
Germany
Orders LinkContactUsLinkComponent
Urea

Urea

5 kg
Catalog # MOLE59164637-5KG
Marketsource
Supplier:   Molekula
Urea
Catalog # MOLE59164637-5KG
Supplier:  Molekula
Marketsource
CAS Number:  57-13-6
Urea
CAS Number:  57-13-6
Price
327,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
  • CAS Number:57-13-6
  • Formula:NH₂CONH₂
  • MDL Number:MFCD00008022
  • Molecular Weight:60.06 g/mol
  • Merck Index:13,09935
  • Boiling Pt:197 °C (1013 hPa)
  • Melting Pt:132.7 °C
  • Density:1.34 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-315-5
  • REACH:01-2119463277-33
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.