Order Entry
Germany
ContactUsLinkComponent
 
Urea 98.0-102.0% (by HPLC), crystalline, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, SAFC®
 :  Merck
 
undefined
Urea 98.0-102.0% (by HPLC), crystalline, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, SAFC®
 :  Merck
 

 

  • Assay (HPLC):
    98.0 - 102.0%
  • Assay (ex N):
    99.0 - 100.5%
  • Assay (ex N, Calculated on Dried Substance):
    99.0 - 101.0%
  • Identity:
    Conforms
  • In Ethanol (96%) Insoluble Parts:
    ≤0.04%
  • Alkalinity:
    Passes Test
  • Chloride (Cl):
    ≤0.007%
  • Sulfate (SO₄):
    ≤0.010%
  • Heavy Metals (as Pb):
    ≤0.0010%
  • NH₄ (Ammonium):
    ≤0.0500%
  • Biuret:
    ≤0.1%
  • Organic Impurities (Related compound A):
    ≤0.1%
  • Organic Impurities (Any Individual Unspecified Impurity):
    ≤0.1%
  • Organic Impurities (Total Impurities):
    ≤2.0%
  • Residual Solvents (ICH Q3C):
    Excluded by Manufacturing Process
  • Sulfated Ash (600 °C):
    ≤0.10%
  • Loss on Drying (105 °C):
    ≤1.0%