Urea, Sigma-Aldrich®
: Merck
: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- :57-13-6
- :NH₂CONH₂
- :MFCD00008022
- :60.06 g/mol
- :13,09935
- :197 °C (1013 hPa)
- :132.7 °C
- :1.34 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :Ambient
- :200-315-5
- :01-2119463277-33
: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- :57-13-6
- :NH₂CONH₂
- :MFCD00008022
- :60.06 g/mol
- :13,09935
- :197 °C (1013 hPa)
- :132.7 °C
- :1.34 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :Ambient
- :200-315-5
- :01-2119463277-33
Product Family Options
- Pack typePk