Urea ≥99.5%, Ultrapure Bioreagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:57-13-6
:
Formula:NH₂CONH₂
MDL Number:MFCD00008022
Merck Index:13,09935
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.34 g/cm³ (20 °C)
:60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
:GHS07
:GHS08
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.5%
- Assay (NH₂CONH₂):98.0 - 102.0 %
- Appearance:Passes Test
- RNase Activity:Passes Test
- DNase Activity (None Detected):Passes Test
- Protease Activity:Passes Test
- Absorbance of a 5M Solution - 260 nm:≤0.050
- Absorbance of a 5M Solution - 280 nm:≤0.030
- Biuret:≤0.01
- Cyanate (CNO):None Detected
- Insoluble Matter:≤0.005 %
- Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Conductivity of 8.5M Solution at 25°C, (µmho/cm):≤30
- Chloride (Cl):≤1 ppm
- Trace Impurities - Copper (Cu):≤1 ppm
- Trace Impurities - Iron (Fe):≤1 ppm
- Trace Impurities - Lead (Pb):≤1 ppm